Assay Details
| Gene Symbol: LOC101747860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300570.2 | XP_015156056.1 | 8 - 9 | 1513 | 61 | |
| XM_015300573.2 | XP_015156059.1 | 8 - 9 | 2865 | 61 | ||
| XM_025145888.1 | XP_025001656.1 | 8 - 9 | 1472 | 61 | ||
| XM_025145889.1 | XP_025001657.1 | 7 - 8 | 1445 | 61 | ||
| XM_025145890.1 | XP_025001658.1 | 8 - 9 | 1433 | 61 | ||
| XM_025145891.1 | XP_025001659.1 | 8 - 9 | 1473 | 61 | ||
| XM_025145892.1 | XP_025001660.1 | 7 - 8 | 1117 | 61 | ||
| XM_025145893.1 | XP_025001661.1 | 7 - 8 | 1486 | 61 | ||
| XM_025145894.1 | XP_025001662.1 | 7 - 8 | 3436 | 61 | ||
| XM_025145895.1 | XP_025001663.1 | 7 - 8 | 2691 | 61 | ||
| XM_025145896.1 | XP_025001664.1 | 4 - 5 | 707 | 61 | ||
More Information
Set Membership: |
|

LOC101747860
Genomic Map