Assay Details
| Gene Symbol: LIPML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234208.4 | XP_001234209.1 | 2 - 3 | 229 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIPML1| Gene Symbol: LIPML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234208.4 | XP_001234209.1 | 2 - 3 | 229 | 106 | |
Set Membership: |
|