Assay Details
| Gene Symbol: LIPML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278700.2 | XP_015134186.1 | 8 - 9 | 1676 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIPML2| Gene Symbol: LIPML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278700.2 | XP_015134186.1 | 8 - 9 | 1676 | 122 | |
Set Membership: |
|