Assay Details
| Gene Symbol: HOXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942724.3 | XP_004942781.1 | 1 - 2 | 4254 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXD1| Gene Symbol: HOXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942724.3 | XP_004942781.1 | 1 - 2 | 4254 | 83 | |
Set Membership: |
|