Assay Details
| Gene Symbol: LOC107050016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145770.1 | XP_025001538.1 | 11 - 12 | 1412 | 91 | |
| XM_025145771.1 | XP_025001539.1 | 12 - 13 | 1297 | 91 | ||
| XM_025145772.1 | XP_025001540.1 | 12 - 13 | 1205 | 91 | ||
More Information
Set Membership: |
|

LOC107050016
Genomic Map