Assay Details
| Gene Symbol: LOC107050016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145770.1 | XP_025001538.1 | 15 - 16 | 1729 | 122 | |
| XM_025145771.1 | XP_025001539.1 | 16 - 17 | 1614 | 122 | ||
| XM_025145772.1 | XP_025001540.1 | 16 - 17 | 1522 | 122 | ||
More Information
Set Membership: |
|

LOC107050016
Genomic Map