Assay Details
| Gene Symbol: LOC107050016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145770.1 | XP_025001538.1 | 5 - 6 | 658 | 60 | |
| XM_025145771.1 | XP_025001539.1 | 6 - 7 | 543 | 60 | ||
| XM_025145772.1 | XP_025001540.1 | 6 - 7 | 451 | 60 | ||
More Information
Set Membership: |
|

LOC107050016
Genomic Map