Assay Details
| Gene Symbol: LOC107050016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145770.1 | XP_025001538.1 | 7 - 8 | 805 | 114 | |
| XM_025145771.1 | XP_025001539.1 | 8 - 9 | 690 | 114 | ||
| XM_025145772.1 | XP_025001540.1 | 8 - 9 | 598 | 114 | ||
More Information
Set Membership: |
|

LOC107050016
Genomic Map