Assay Details
| Gene Symbol: LOC101749023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278810.2 | XP_015134296.2 | 5 - 6 | 1169 | 107 | |
| XM_015278817.2 | XP_015134303.2 | 5 - 6 | 1235 | 107 | ||
| XM_015278822.2 | XP_015134308.2 | 5 - 6 | 1876 | 107 | ||
| XM_015278832.2 | XP_015134318.2 | 4 - 5 | 1011 | 107 | ||
| XM_015278845.2 | XP_015134331.2 | 2 - 3 | 739 | 107 | ||
| XM_015278851.2 | XP_015134337.2 | 3 - 4 | 841 | 107 | ||
| XM_015278861.2 | XP_015134347.2 | 2 - 3 | 652 | 107 | ||
| XM_015278875.2 | XP_015134361.2 | 2 - 3 | 664 | 107 | ||
| XM_025146839.1 | XP_025002607.1 | 4 - 5 | 999 | 107 | ||
More Information
Set Membership: |
|

LOC101749023
Genomic Map