Assay Details
| Gene Symbol: LOC107049603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015273318.2 | XP_015128804.2 | 1 - 2 | 1747 | 90 | |
| XM_025144513.1 | XP_025000281.1 | 1 - 2 | 1745 | 90 | ||
More Information
Set Membership: |
|

LOC107049603
Genomic Map