Assay Details
| Gene Symbol: LOC101751434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025143996.1 | XP_024999764.1 | 2 - 3 | 643 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101751434| Gene Symbol: LOC101751434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025143996.1 | XP_024999764.1 | 2 - 3 | 643 | 95 | |
Set Membership: |
|