Assay Details
| Gene Symbol: MFSD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935790.3 | XP_004935847.1 | 10 - 11 | 2123 | 60 | |
| XM_426214.6 | XP_426214.4 | 10 - 11 | 1248 | 60 | ||
More Information
Set Membership: |
|

MFSD2B
Genomic Map