Assay Details
| Gene Symbol: CA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153922.1 | XP_025009690.1 | 4 - 5 | 459 | 73 | |
| XM_413756.5 | XP_413756.3 | 6 - 7 | 755 | 73 | ||
More Information
Set Membership: |
|

CA12
Genomic Map