Assay Details
| Gene Symbol: CERS4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144309.1 | XP_025000077.1 | 9 - 10 | 2873 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CERS4L| Gene Symbol: CERS4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144309.1 | XP_025000077.1 | 9 - 10 | 2873 | 80 | |
Set Membership: |
|