Assay Details
| Gene Symbol: ATP5SL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015274099.2 | XP_015129585.2 | 5 - 6 | 965 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP5SL| Gene Symbol: ATP5SL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015274099.2 | XP_015129585.2 | 5 - 6 | 965 | 99 | |
Set Membership: |
|