Assay Details
| Gene Symbol: CBX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288679.2 | XP_015144165.2 | 3 - 4 | 261 | 112 | |
| XM_015288685.2 | XP_015144171.2 | 3 - 4 | 261 | 112 | ||
More Information
Set Membership: |
|

CBX6
Genomic Map