Assay Details
| Gene Symbol: KCNN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300089.2 | XP_015155575.2 | 2 - 3 | 507 | 85 | |
| XM_015300090.2 | XP_015155576.2 | 2 - 3 | 504 | 85 | ||
| XM_015300091.2 | XP_015155577.2 | 2 - 3 | 497 | 85 | ||
| XM_025144393.1 | XP_025000161.1 | 2 - 3 | 500 | 85 | ||
| XM_025144394.1 | XP_025000162.1 | 2 - 3 | 503 | 85 | ||
More Information
Set Membership: |
|

KCNN1
Genomic Map