Assay Details
| Gene Symbol: TPM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300190.1 | XP_015155676.1 | 1 - 2 | 271 | 107 | |
| XM_015300191.2 | XP_015155677.1 | 1 - 2 | 367 | 107 | ||
| XM_025144432.1 | XP_025000200.1 | 9 - 10 | 11501 | 107 | ||
More Information
Set Membership: |
|

TPM4
Genomic Map