Assay Details
| Gene Symbol: LOC107051108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145714.1 | XP_025001482.1 | 1 - 2 | 247 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107051108| Gene Symbol: LOC107051108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145714.1 | XP_025001482.1 | 1 - 2 | 247 | 99 | |
Set Membership: |
|