Assay Details
| Gene Symbol: MXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293858.2 | XP_015149344.2 | 10 - 11 | 1399 | 74 | |
| XM_015293859.2 | XP_015149345.2 | 10 - 11 | 1399 | 74 | ||
More Information
Set Membership: |
|

MXD3
Genomic Map