Assay Details
| Gene Symbol: PLA2G4EL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234823.5 | XP_001234824.3 | 6 - 7 | 608 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G4EL4| Gene Symbol: PLA2G4EL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234823.5 | XP_001234824.3 | 6 - 7 | 608 | 70 | |
Set Membership: |
|