Assay Details
| Gene Symbol: GPR153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297062.2 | XP_015152548.1 | 5 - 6 | 1414 | 93 | |
| XM_015297063.2 | XP_015152549.1 | 5 - 6 | 1932 | 93 | ||
More Information
Set Membership: |
|

GPR153
Genomic Map