Assay Details
| Gene Symbol: WFIKKN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155291.1 | XP_025011059.1 | 1 - 2 | 1873 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WFIKKN1| Gene Symbol: WFIKKN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155291.1 | XP_025011059.1 | 1 - 2 | 1873 | 117 | |
Set Membership: |
|