Assay Details
| Gene Symbol: LOC101751912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145921.1 | XP_025001689.1 | 1 - 2 | 171 | 100 | |
| XM_025145922.1 | XP_025001690.1 | 1 - 2 | 172 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025145921.1, XM_025145922.1 transcripts.
Set Membership: |
|

LOC101751912
Genomic Map