Assay Details
| Gene Symbol: LOC101751912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145921.1 | XP_025001689.1 | 3 - 4 | 658 | 69 | |
| XM_025145922.1 | XP_025001690.1 | 3 - 4 | 659 | 69 | ||
More Information
Set Membership: |
|

LOC101751912
Genomic Map