Assay Details
| Gene Symbol: LOC101751912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145921.1 | XP_025001689.1 | 7 - 8 | 1102 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101751912| Gene Symbol: LOC101751912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145921.1 | XP_025001689.1 | 7 - 8 | 1102 | 129 | |
Set Membership: |
|