Assay Details
Gene Symbol: CCDC110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004935976.3 | XP_004936033.2 | 2 - 3 | 467 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
