Assay Details
| Gene Symbol: LOC424300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025152679.1 | XP_025008447.1 | 7 - 8 | 5044 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC424300| Gene Symbol: LOC424300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025152679.1 | XP_025008447.1 | 7 - 8 | 5044 | 109 | |
Set Membership: |
|