Assay Details
| Gene Symbol: ERI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155185.1 | XP_025010953.1 | 3 - 4 | 399 | 75 | |
| XM_424602.6 | XP_424602.4 | 3 - 4 | 399 | 75 | ||
More Information
Set Membership: |
|

ERI2
Genomic Map