Assay Details
| Gene Symbol: LOC416696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286002.2 | XP_015141488.2 | 39 - 40 | 4901 | 85 | |
| XM_025153151.1 | XP_025008919.1 | 39 - 40 | 4902 | 85 | ||
| XM_025153153.1 | XP_025008921.1 | 39 - 40 | 4901 | 85 | ||
More Information
Set Membership: |
|

LOC416696
Genomic Map