Assay Details
| Gene Symbol: LOC416696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286002.2 | XP_015141488.2 | 63 - 64 | 6370 | 65 | |
| XM_025153151.1 | XP_025008919.1 | 63 - 64 | 6371 | 65 | ||
| XM_025153153.1 | XP_025008921.1 | 62 - 63 | 6334 | 65 | ||
More Information
Set Membership: |
|

LOC416696
Genomic Map