Assay Details
Gene Symbol: FRMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015278445.2 | XP_015133931.1 | 3 - 4 | 1991 | 108 | |
XM_015278448.2 | XP_015133934.1 | 2 - 3 | 789 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015278448.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
