Assay Details
Gene Symbol: HTR1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284707.2 | XP_015140193.1 | 1 - 2 | 198 | 74 | |
XM_015284708.2 | XP_015140194.1 | 1 - 2 | 198 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015284707.2, XM_015284708.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
