Assay Details
Gene Symbol: HTR1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284707.2 | XP_015140193.1 | 3 - 4 | 1177 | 145 | |
XM_025149278.1 | XP_025005046.1 | 1 - 2 | 1029 | 145 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015284707.2, XM_025149278.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
