Assay Details
| Gene Symbol: SLC2A4RG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142444.1 | XP_024998212.1 | 4 - 5 | 2283 | 83 | |
| XM_025142445.1 | XP_024998213.1 | 4 - 5 | 2280 | 83 | ||
More Information
Set Membership: |
|

SLC2A4RG
Genomic Map