Assay Details
| Gene Symbol: PKDREJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015290757.2 | XP_015146243.2 | 1 | 7179 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PKDREJ| Gene Symbol: PKDREJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015290757.2 | XP_015146243.2 | 1 | 7179 | 101 | |
Set Membership: |
|