Assay Details
| Gene Symbol: ZNF653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015302690.2 | XP_015158176.1 | 6 - 7 | 1353 | 58 | |
| XM_015302691.2 | XP_015158177.1 | 6 - 7 | 1356 | 58 | ||
| XM_015302692.2 | XP_015158178.1 | 6 - 7 | 1355 | 58 | ||
More Information
Set Membership: |
|

ZNF653
Genomic Map