Assay Details
| Gene Symbol: LOC107050018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015273823.2 | XP_015129309.2 | 6 - 7 | 683 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107050018| Gene Symbol: LOC107050018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015273823.2 | XP_015129309.2 | 6 - 7 | 683 | 71 | |
Set Membership: |
|