Assay Details
Gene Symbol: SYDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025145753.1 | XP_025001521.1 | 3 - 4 | 1965 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
