Assay Details
| Gene Symbol: CCDC160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298185.2 | XP_015153671.1 | 12 - 13 | 1393 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC160| Gene Symbol: CCDC160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298185.2 | XP_015153671.1 | 12 - 13 | 1393 | 105 | |
Set Membership: |
|