Assay Details
| Gene Symbol: CCDC160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298185.2 | XP_015153671.1 | 6 - 7 | 613 | 76 | |
| XM_015298186.2 | XP_015153672.1 | 6 - 7 | 614 | 76 | ||
More Information
Set Membership: |
|

CCDC160
Genomic Map