Assay Details
| Gene Symbol: LOC112531518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025146689.1 | XP_025002457.1 | 1 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025146656.1, XM_025146657.1, XM_025146658.1, XM_025146659.1, XM_025146660.1, XM_025146661.1, XM_025146662.1, XM_025146663.1, XM_025146664.1, XM_025146665.1, XM_025146666.1, XM_025146667.1, XM_025146668.1, XM_025146669.1, XM_025146670.1, XM_025146671.1, XM_025146672.1, XM_025146673.1, XM_025146674.1, XM_025146675.1, XM_025146677.1, XM_025146678.1, XM_025146683.1, XM_025146686.1, XM_025146687.1, XM_025146689.1 transcripts.
Set Membership: |
|

LOC776067
Genomic Map