Assay Details
| Gene Symbol: GJA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025143016.1 | XP_024998784.1 | 2 | 4395 | 59 | |
| XM_025143017.1 | XP_024998785.1 | 2 | 4395 | 59 | ||
More Information
Set Membership: |
|

GJA9
Genomic Map