Assay Details
| Gene Symbol: LOC101747373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025151820.1 | XP_025007588.1 | 6 | 6475 | 132 | |
| XM_025151821.1 | XP_025007589.1 | 6 | 6458 | 132 | ||
| Gene Symbol: ZNF251L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153971.1 | XP_025009739.1 | 1 | 2917 | 132 | |
More Information
Set Membership: |
|

LOC101747373
Genomic Map