Assay Details
| Gene Symbol: LRRC3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299547.2 | XP_015155033.1 | 2 | 1814 | 85 | |
| XM_025144062.1 | XP_024999830.1 | 2 | 1879 | 85 | ||
More Information
Set Membership: |
|

LRRC3C
Genomic Map