Assay Details
| Gene Symbol: LOC107055860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147745.1 | XP_025003513.1 | 1 | 484 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025147745.1 transcripts.
Set Membership: |
|

LOC107055860
Genomic Map