Assay Details
| Gene Symbol: LOC107055859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025148005.1 | XP_025003773.1 | 5 | 1440 | 92 | |
| XM_025148006.1 | XP_025003774.1 | 3 | 982 | 92 | ||
| XM_025148007.1 | XP_025003775.1 | 2 | 1085 | 92 | ||
| XM_025148008.1 | XP_025003776.1 | 5 | 1440 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025148005.1, XM_025148006.1, XM_025148007.1, XM_025148008.1 transcripts.
Set Membership: |
|

LOC107055859
Genomic Map