Assay Details
| Gene Symbol: GPR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938778.3 | XP_004938835.1 | 2 | 1204 | 80 | |
| XM_025146842.1 | XP_025002610.1 | 2 | 1288 | 80 | ||
More Information
Set Membership: |
|

GPR12
Genomic Map