Assay Details
| Gene Symbol: SRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249730.1 | NP_001236659.1 | 1 | 451 | 91 | |
| GenBank mRNA | AB000130.1 | - | 1 | 451 | 91 | |
| AK245187.1 | - | 1 | 543 | 91 | ||
More Information
Set Membership: |
|

SRC2
Genomic Map