Assay Details
| Gene Symbol: MYB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255462.2 | NP_001242391.1 | 2 - 3 | 357 | 72 | |
| GenBank mRNA | AB029269.1 | - | 1 - 2 | 93 | 72 | |
| BT097379.1 | - | 2 - 3 | 359 | 72 | ||
More Information
Set Membership: |
|

MYB12
Genomic Map